4-(Trifluoromethylthio)phenol ≥98.0%

Supplier: TCI America

Synonyms: 4-[(Trifluoromethyl)sulphanyl]phenol

T2458-5G T2458-25G
TCT2458-5GEA 34.6 USD
TCT2458-5G TCT2458-25G
4-(Trifluoromethylthio)phenol ≥98.0%
4-(Trifluoromethylthio)phenol

CAS Number: 461-84-7
MDL Number: MFCD00036035
Molecular Formula: C7H5F3OS
Molecular Weight: 194.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 78
Melting point (°C): 60

Formula: C₇H₅F₃OS
Boiling Pt: 78 °C
Melting Pt: 60 °C
MDL Number: MFCD00036035
CAS Number: 461-84-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e8c5ec0e2f8f0dc3c9cf6e2396dab8ac07bc594&vid=4e8c5ec0e2f8f0dc3c9cf6e2396dab8ac07bc594&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR