4-(Trifluoromethoxy)phenylhydrazine hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
(4-(Trifluoromethoxy)phenyl)hydrazine hydrochloride
T2415-1G
T2415-5G
TCT2415-1GEA
42
USD
TCT2415-1G
TCT2415-5G
4-(Trifluoromethoxy)phenylhydrazine hydrochloride ≥98.0%
4-(Trifluoromethoxy)phenylhydrazine hydrochloride
CAS Number: 133115-72-7
MDL Number: MFCD00053033
Molecular Formula: C7H7F3N2O
Molecular Weight: 228.60
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 230
Formula:
C₇H₈ClF₃N₂O Melting Pt: 230 °C |
MDL Number:
MFCD00053033 CAS Number: 133115-72-7 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...