2-(Trifluoromethyl)phenylacetonitrile ≥98.0%

Supplier: TCI America

Synonyms: 2-(Trifluoromethyl)benzyl cyanide, 1-(Cyanmethyl)-2-(trifluormethyl)benzene

T2366-5G T2366-25G
TCT2366-5GEA 35.6 USD
TCT2366-5G TCT2366-25G
2-(Trifluoromethyl)phenylacetonitrile ≥98.0%
2-(Trifluoromethyl)phenylacetonitrile

CAS Number: 3038-47-9
MDL Number: MFCD00001903
Molecular Formula: C9H6F3N
Molecular Weight: 185.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 92
Flash Point (°C): 89
Freezing point (°C): 31


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-(Trifluoromethyl)benzyl Cyanide

Formula: C₉H₆F₃N
MW: 185.15 g/mol
Boiling Pt: 92 °C
Flash Pt: 89 °C
MDL Number: MFCD00001903
CAS Number: 3038-47-9
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80012c18a2d06d799ec0ca0cf7579f0fc3891fd2&vid=80012c18a2d06d799ec0ca0cf7579f0fc3891fd2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR