3,4,5-Trifluoroaniline ≥98.0%
Supplier: TCI America
T2355-5G
T2355-1G
TCT2355-5GEA
84
USD
TCT2355-5G
TCT2355-1G
3,4,5-Trifluoroaniline ≥98.0%
3,4,5-Trifluoroaniline
CAS Number: 163733-96-8
MDL Number: MFCD00083522
Molecular Formula: C6H4F3N
Molecular Weight: 147.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 64
Formula:
C₆H₄F₃N MW: 147.1 g/mol Melting Pt: 64 °C |
MDL Number:
MFCD00083522 CAS Number: 163733-96-8 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...