2,5-Thiophenedicarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Thiophene-2,5-dicarboxylic acid

T2347-25G T2347-5G
TCT2347-25GEA 130.8 USD
TCT2347-25G TCT2347-5G
2,5-Thiophenedicarboxylic acid ≥98.0% (by GC, titration analysis)
2,5-Thiophenedicarboxylic acid

CAS Number: 4282-31-9
MDL Number: MFCD00016896
Molecular Formula: C6H4O4S
Molecular Weight: 172.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 359

Formula: C₆H₄O₄S
MW: 172.16 g/mol
Melting Pt: 359 °C
MDL Number: MFCD00016896
CAS Number: 4282-31-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0db2882f89cdd4fe46804d532775aeb2ea643fd6&vid=0db2882f89cdd4fe46804d532775aeb2ea643fd6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR