3-(Trifluoromethoxy)aniline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
3-Trifluoromethoxyaniline
T2255-1G
T2255-5G
TCT2255-1GEA
30.53
USD
TCT2255-1G
TCT2255-5G
3-(Trifluoromethoxy)aniline ≥98.0% (by GC, titration analysis)
3-(Trifluoromethoxy)aniline
CAS Number: 1535-73-5
Molecular Formula: C7H6F3NO
Molecular Weight: 177.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 70
Flash Point (°C): 85
Specific Gravity (20/20): 1.33
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha-Trifluoro-m-anisidine
Formula:
C₇H₆F₃NO MW: 177.13 g/mol Boiling Pt: 70 °C Density: 1.33 (20 °C) Flash Pt: 85 °C |
MDL Number:
MFCD00041511 CAS Number: 1535-73-5 UN: 2810 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...