2,3,4,5-Tetrafluorobenzonitrile ≥96.0%

Supplier: TCI America

T1882-1G T1882-5G
TCT1882-1GEA 49 USD
TCT1882-1G TCT1882-5G
2,3,4,5-Tetrafluorobenzonitrile ≥96.0%
2,3,4,5-Tetrafluorobenzonitrile

CAS Number: 16582-93-7
MDL Number: MFCD00040187
Molecular Formula: C7HF4N
Molecular Weight: 175.09
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 165
Flash Point (°C): 70
Specific Gravity (20/20): 1.48

Formula: C₇HF₄N
MW: 175.09 g/mol
Boiling Pt: 165 °C
Density: 1.48 (20 °C)
Flash Pt: 70 °C
MDL Number: MFCD00040187
CAS Number: 16582-93-7
UN: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a22248b3ba29394a432c2b27943c3afe5a2cb654&vid=a22248b3ba29394a432c2b27943c3afe5a2cb654&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR