1,2,3-Triazole ≥98.0%

Supplier: TCI America

Synonyms: 1,2,3-1H-Triazole, 1H-1,2,3-Triazole

T1880-25G T1880-5G
TCT1880-25GEA 404 USD
TCT1880-25G TCT1880-5G
1,2,3-Triazole ≥98.0%
1,2,3-Triazole

CAS Number: 288-36-8
MDL Number: MFCD00014490
Molecular Formula: C2H3N3
Molecular Weight: 69.07
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Flash Point (°C): 107
Freezing point (°C): 22

Formula: C₂H₃N₃
Flash Pt: 107 °C
MDL Number: MFCD00014490
CAS Number: 288-36-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3dd571c5f8a36d4e4c17a6e31754d54fe4d283fa&vid=3dd571c5f8a36d4e4c17a6e31754d54fe4d283fa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR