Methyl trifluoropyruvate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 3,3,3-trifluoropyruvate, Methyl 3,3,3-trifluoro-2-oxopropanoate

T1785-1G T1785-5G
TCT1785-001GEA 18 USD
TCT1785-001G TCT1785-005G
Methyl trifluoropyruvate ≥98.0%
Methyl trifluoropyruvate

CAS Number: 13089-11-7
MDL Number: MFCD00114936
Molecular Formula: C4H3F3O3
Molecular Weight: 156.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Slightly Pale Yellow
Boiling point (°C): 86
Flash Point (°C): 26
Specific Gravity (20/20): 1.39


Other Product Names / Synonyms: Trifluoropyruvic Acid Methyl Ester

Formula: C₄H₃F₃O₃
MW: 156.06 g/mol
Boiling Pt: 86 °C
Density: 1.39 (20 °C)
Flash Pt: 26 °C
MDL Number: MFCD00114936
CAS Number: 13089-11-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6e49ccb5083916e3d68d67b9f97df7db1fe91a5&vid=e6e49ccb5083916e3d68d67b9f97df7db1fe91a5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR