Tetrabutylammonium difluorotriphenylstannate ≥97.0% (by titrimetric analysis)
Supplier: TCI America
T1592-1G
T1592-5G
TCT1592-001GEA
76
USD
TCT1592-001G
TCT1592-5G
Tetrabutylammonium difluorotriphenylstannate ≥97.0% (by titrimetric analysis)
Tetrabutylammonium difluorotriphenylstannate
CAS Number: 139353-88-1
MDL Number: MFCD00192465
Molecular Formula: C34H51F2NSn
Molecular Weight: 630.50
Purity/Analysis Method:
Formula:
C₃₄H₅₁F₂NSn Melting Pt: 193 °C |
MDL Number:
MFCD00192465 CAS Number: 139353-88-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...