Ethyl trifluoropyruvate ≥98.0%
Supplier: TCI America
Synonyms:
Ethyl 3,3,3-trifluoro-2-oxopropanoate, Ethyl 3,3,3-trifluoropyruvate
T1434-5G
T1434-25G
TCT1434-005GEA
48
USD
TCT1434-005G
TCT1434-25G
Ethyl trifluoropyruvate ≥98.0%
Ethyl trifluoropyruvate
CAS Number: 13081-18-0
MDL Number: MFCD00114935
Molecular Formula: C5H5F3O3
Molecular Weight: 170.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 98
Flash Point (°C): 31
Specific Gravity (20/20): 1.28
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Trifluoropyruvic Acid Ethyl Ester
Formula:
C₅H₅F₃O₃ MW: 170.09 Boiling Pt: 98 °C Density: 1.28 (20 °C) Flash Pt: 31 °C |
MDL Number:
MFCD00114935 CAS Number: 13081-18-0 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...