3,5,6-Trichlorosalicylic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
3,5,6-Trichlorosalicylic Acid
T1406-500G
T1406-25G
TCT1406-500GEA
320
USD
TCT1406-500G
TCT1406-025G
3,5,6-Trichlorosalicylic acid ≥97.0% (by GC, titration analysis)
3,5,6-Trichlorosalicylic acid
CAS Number: 40932-60-3
MDL Number: MFCD00075245
Molecular Formula: C7H3Cl3O3
Molecular Weight: 241.45
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 208
Other Product Names / Synonyms: 3,5,6-Trichloro-2-hydroxybenzoic Acid
Formula:
C₇H₃Cl₃O₃ MW: 241.46 g/mol Melting Pt: 208 °C |
MDL Number:
MFCD00075245 CAS Number: 40932-60-3 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...