Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')europium(III) ≥95.0%
Supplier: TCI America
Synonyms:
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)europium(III), Eu(TMHD)3
T1265-1G
T1265-5G
TCT1265-001GEA
51
USD
TCT1265-001G
TCT1265-5G
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')europium(III) ≥95.0%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')europium(III)
[NMR Shift Reagent]
CAS Number: 15522-71-1
MDL Number: MFCD00000019
Molecular Formula: C33H57EuO6
Molecular Weight: 701.78
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 190
Product Categories: Analytical Science, Shift Reagents
Other Product Names / Synonyms: Eu(dpm)3, Europium(III) Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)
Formula:
C₃₃H₅₇EuO₆ Melting Pt: 190 °C |
MDL Number:
MFCD00000019 CAS Number: 15522-71-1 |
|
Specification Test Results
Purity/Analysis Method | >95.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...