2-Thiopheneacrylic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (E)-3-(Thiophen-2-yl)acrylic acid, 3-(2-Thienyl)acrylic Acid

T1138-25G T1138-5G
TCT1138-025GEA 254.2 USD
TCT1138-025G TCT1138-005G
2-Thiopheneacrylic acid ≥98.0% (by titrimetric analysis)
2-Thiopheneacrylic acid

CAS Number: 15690-25-2
MDL Number: MFCD00005455
Molecular Formula: C7H6O2S
Molecular Weight: 154.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 146


Other Product Names / Synonyms: 3-(2-Thienyl)acrylic Acid

Formula: C₇H₆O₂S
MW: 154.18 g/mol
Melting Pt: 146 °C
MDL Number: MFCD00005455
CAS Number: 15690-25-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=260a9bfd5f44ee972bf8417f47d28168e087b8c5&vid=260a9bfd5f44ee972bf8417f47d28168e087b8c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR