Trimethyl orthovalerate

Supplier: TCI America

Synonyms: 1,1,1-Trimethoxypentane, Orthovaleric Acid Trimethyl Ester

T0981-25ML T0981-250ML
TCT0981-025MLEA 81.6 USD
TCT0981-025ML TCT0981-250ML
Trimethyl orthovalerate
Trimethyl orthovalerate

CAS Number: 13820-09-2
MDL Number: MFCD00008481
Molecular Formula: C8H18O3
Molecular Weight: 162.23
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 164
Flash Point (°C): 55
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: 1,1,1-Trimethoxypentane, Orthovaleric Acid Trimethyl Ester

Formula: C₈H₁₈O₃
MW: 162.23 g/mol
Boiling Pt: 164 °C
Density: 0.92 (20 °C)
Flash Pt: 55 °C
MDL Number: MFCD00008481
CAS Number: 13820-09-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=165c5001ed7355e2e1ef2f87e66ae1fec0ccdb04&vid=165c5001ed7355e2e1ef2f87e66ae1fec0ccdb04&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR