Tetrabutylammonium hydroxide 10% in water

Supplier: TCI America

T0955-500ML T0955-25ML T0955-100ML
TCT0955-500MLEA 217.19 USD
TCT0955-500ML TCT0955-25ML TCT0955-100ML
Tetrabutylammonium hydroxide 10% in water
Tetrabutylammonium hydroxide

CAS Number: 2052-49-5
MDL Number: MFCD00009425
Molecular Formula: C16H37NO
Molecular Weight: 259.48
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.00

Formula: C₁₆H₃₇NO
MW: 259.48 g/mol
Density: 1.00 (20 °C)
MDL Number: MFCD00009425
CAS Number: 2052-49-5
UN: 3263
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ceec2fc79a22dbb6e500efa0b64ebd2efdbbc4d&vid=c014fc5494b0db84fd4af0091e75c6041675b014&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR