Hexaketocyclohexane ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Hexaoxocyclohexane, Triquinoyl, Cyclohexanehexone

T0876-1G T0876-5G
TCT0876-1GEA 32 USD
TCT0876-1G TCT0876-005G
Hexaketocyclohexane ≥95.0% (by titrimetric analysis)
Hexaketocyclohexane

CAS Number: 527-31-1
MDL Number: MFCD00149075
Molecular Formula: C6O6
Molecular Weight: 168.06
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White
Melting point (°C): 93
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Cyclohexanehexone, Hexaoxocyclohexane

Formula: C₆O₆
MW: 168.06 g/mol
Melting Pt: 93 °C
MDL Number: MFCD00001652
CAS Number: 527-31-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=941b2c326acfd48165b5ed7982adbd80150f5955&vid=941b2c326acfd48165b5ed7982adbd80150f5955&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR