DL-Tartaric acid ≥99.0% (by titrimetric analysis)
Supplier: TCI America
T0001-500G
T0001-25G
TCT0001-500GEA
75
USD
TCT0001-500G
TCT0001-025G
DL-Tartaric acid ≥99.0% (by titrimetric analysis)
DL-Tartaric acid
CAS Number: 133-37-9
MDL Number: MFCD00151254
Molecular Formula: C4H6O6
Molecular Weight: 150.09
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Color: White
Melting point (°C): 199
Flash Point (°C): 210
Other Product Names / Synonyms: Racemic Acid
Formula:
C₄H₆O₆ MW: 150.09 g/mol Melting Pt: 199 °C Flash Pt: 210 °C |
MDL Number:
MFCD00151254 CAS Number: 133-37-9 |
Specification Test Results
Purity/Analysis Method | >99.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...