3-Maleimidopropionic acid N-hydroxysuccinimide ester ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: N-(3-Maleimidopropionyloxy)succinimide, 2,5-Dioxopyrrolidin-1-yl 3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoate, N-Succinimidyl 3-Maleimidopropionat, 3-Maleimidopropionic Acid N-Succinimidyl Ester, N-Succinimidyl 3-maleimidopropionate, 3-(Maleimido)propionic acid N-hydroxysuccinimide ester, N-β-maleimidopropyl-oxysuccinimide ester, BMPS

S0427-1G S0427-100MG
TCS0427-001GEA 256.46 USD
TCS0427-001G TCS0427-100MG
3-Maleimidopropionic acid N-hydroxysuccinimide ester ≥98.0% (by HPLC, total nitrogen)
BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester)

[Cross-linking Reagent]
CAS Number: 55750-62-4
MDL Number: MFCD00043141
Molecular Formula: C11H10N2O6
Molecular Weight: 266.21
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 168
Storage Temperature: 0-10°C


Product Categories: Bioscience, Crosslinkers
Other Product Names / Synonyms: 3-Maleimidopropionic Acid N-Succinimidyl Ester, N-(3-Maleimidopropionyloxy)succinimide, BMPS

Formula: C₁₁H₁₀N₂O₆
MW: 266.21 g/mol
Melting Pt: 168 °C
MDL Number: MFCD00043141
CAS Number: 55750-62-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49abc4558a5a8f49f9eb32662a511eddc85c8a0c&vid=49abc4558a5a8f49f9eb32662a511eddc85c8a0c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR