Retinoic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
all-trans-Retinoic acid, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid, ATRA, Vitamin A acid, Tretinoin
CAS Number: 302-79-4
MDL Number: MFCD00001551
Molecular Formula: C20H28O2
Molecular Weight: 300.44
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 180
Product Categories: Bioscience, Regenerative Medicine Research Reagents, Antitumor Ingredients, Vitamins, Terpenes (Others)
Other Product Names / Synonyms: Vitamin A Acid, all-trans-Retinoic Acid, Tretinoin, all-trans-3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexen-1-yl)-2,4,6,8-nonatetraenoic Acid
Formula:
C₂₀H₂₈O₂ MW: 300.44 g/mol Melting Pt: 180 °C |
MDL Number:
MFCD00001551 CAS Number: 302-79-4 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...