2-Quinolinecarbaldehyde ≥97.0%

Supplier: TCI America

Synonyms: Quinoline-2-carboxaldehyde, Quinoline-2-carbaldehyde, 2-Quinolinecarboxaldehyde

Q0064-1G Q0064-5G
TCQ0064-1GEA 39 USD
TCQ0064-1G TCQ0064-5G
2-Quinolinecarbaldehyde ≥97.0%
2-Quinolinecarbaldehyde

CAS Number: 5470-96-2
MDL Number: MFCD00075032
Molecular Formula: C10H7NO
Molecular Weight: 157.17
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 70
Flash Point (°C): 112


Other Product Names / Synonyms: 2-Formylquinoline

Formula: C₁₀H₇NO
MW: 157.17 g/mol
Melting Pt: 70 °C
Flash Pt: 112 °C
MDL Number: MFCD00075032
CAS Number: 5470-96-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27935059d35a5755f9aad674e6fa0ec56e51d188&vid=3b48304115d239babed901f6eaf23510fb3bf60d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR