4-Pyridineboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 4-Pyridinylboronic acid, Pyridine-4-boronic acid, Pyridin-4-ylboronic acid

P1594-5G P1594-1G
TCP1594-5GEA 67 USD
TCP1594-5G TCP1594-1G
4-Pyridineboronic acid (contains varying amounts of Anhydride)
4-Pyridineboronic acid

CAS Number: 1692-15-5
MDL Number: MFCD01074545
Molecular Formula: C5H6BNO2
Molecular Weight: 122.92
Form: Crystal
Color: White


Other Product Names / Synonyms: Pyridine-4-boronic Acid

Formula: C₅H₆BNO₂
MW: 122.92 g/mol (anhy)
MDL Number: MFCD01074545
CAS Number: 1692-15-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8fd8712a8c1db8667d767645fb6cff10cfb68db&vid=c8fd8712a8c1db8667d767645fb6cff10cfb68db&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR