7-Ethoxy-7-oxoheptanoic acid ≥98.0%
Supplier: TCI America
Synonyms:
Monoethyl pimelate, Pimelic acid monoethyl ester, Ethyl yydrogen pimelate
P1244-5G
P1244-25G
TCP1244-005GEA
64.9
USD
TCP1244-005G
TCP1244-025G
7-Ethoxy-7-oxoheptanoic acid ≥98.0%
7-Ethoxy-7-oxoheptanoic acid
CAS Number: 33018-91-6
MDL Number: MFCD00801202
Molecular Formula: C9H16O4
Molecular Weight: 188.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 162
Specific Gravity (20/20): 1.06
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Ethyl Hydrogen Pimelate, Pimelic Acid Monoethyl Ester
Formula:
C₉H₁₆O₄ MW: 188.22 g/mol Boiling Pt: 162 °C Density: 1.06 (20 °C) |
MDL Number:
MFCD00014383 CAS Number: 33018-91-6 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...