(Pentafluorophenyl)diphenylphosphine ≥93.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Diphenyl(pentafluorophenyl)phosphine
P1176-1G
P1176-5G
TCP1176-001GEA
152
USD
TCP1176-001G
TCP1176-005G
(Pentafluorophenyl)diphenylphosphine ≥93.0% (by titrimetric analysis)
(Pentafluorophenyl)diphenylphosphine
CAS Number: 5525-95-1
MDL Number: MFCD00000290
Molecular Formula: C18H10F5P
Molecular Weight: 352.24
Purity/Analysis Method: >93.0% (T)
Form: Crystal
Color: White
Melting point (°C): 70
Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: Diphenyl(pentafluorophenyl)phosphine
Formula:
C₁₈H₁₀F₅P MW: 352.24 g/mol Melting Pt: 70 °C |
MDL Number:
MFCD00000290 CAS Number: 5525-95-1 |
Specification Test Results
Purity/Analysis Method | >93.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...