(±)-Propranolol hydrochloride 99,0

Supplier: TCI America

Synonyms: 1-(Isopropylamino)-3-(1-naphthyloxy)-2-propanol hydrochloride, Propranolol hydrochloride, DL-Propranolol hydrochloride, (±)-1-Isopropylamino-3-(1-naphthyloxy)-2-propanol hydrochloride

P0995-250G P0995-25G
TCP0995-250GEA 736 USD
TCP0995-250G TCP0995-025G
(±)-Propranolol hydrochloride 99,0
(±)-Propranolol hydrochloride

CAS Number: 318-98-9
MDL Number: MFCD00012558
Molecular Formula: C16H21NO2
Molecular Weight: 295.81
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 166


Product Categories: Bioscience, Reagents for Life-style related Disease Models, Antihypertensive Ingredients
Other Product Names / Synonyms: 1-(Isopropylamino)-3-(1-naphthyloxy)-2-propanol Hydrochloride

Formula: C₁₆H₂₁NO₂
MW: 295.81 g/mol
Melting Pt: 166 °C
MDL Number: MFCD00012558
CAS Number: 318-98-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca65abc1908ab66c6dd01bd94658eedb35e2794f&vid=ca65abc1908ab66c6dd01bd94658eedb35e2794f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR