Vinyl propionate ≥98.0% stabilized

Supplier: TCI America

P0613-25ML P0613-500ML
TCP0613-025MLEA 23 USD
TCP0613-025ML TCP0613-500ML
Vinyl propionate ≥98.0% stabilized
Vinyl propionate

(stabilized with MEHQ)
CAS Number: 105-38-4
MDL Number: MFCD00027004
Molecular Formula: C5H8O2
Molecular Weight: 100.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 95
Melting point (°C): -80
Flash Point (°C): 8
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: Propionic Acid Vinyl Ester

Formula: C₅H₈O₂
Boiling Pt: 95 °C
Melting Pt: –80 °C
Density: 0.92 (20 °C)
Flash Pt: 8 °C
MDL Number: MFCD00027004
CAS Number: 105-38-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8beed8b0fa243ef36a83209dbb726c20237c93aa&vid=8beed8b0fa243ef36a83209dbb726c20237c93aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR