Pivalonitrile ≥98.0%
Supplier: TCI America
P0463-25ML
P0463-100ML
TCP0463-25MLEA
66.37
USD
TCP0463-25ML
TCP0463-100ML
Pivalonitrile ≥98.0%
Pivalonitrile
CAS Number: 630-18-2
MDL Number: MFCD00001847
Molecular Formula: C5H9N
Molecular Weight: 83.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 105
Flash Point (°C): 21
Freezing point (°C): 15
Specific Gravity (20/20): 0.76
Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: 2,2-Dimethylpropionitrile, Trimethylacetonitrile
Formula:
C₅H₉N Boiling Pt: 105 °C Density: 0.76 (20 °C) Flash Pt: 21 °C |
MDL Number:
MFCD00001847 CAS Number: 630-18-2 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...