4-(Isopropylamino)diphenylamine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
N1-Isopropyl-N4-phenylbenzene-1,4-diamine, N-Isopropyl-N'-phenyl-p-phenylenediamine
P0327-25G
P0327-500G
TCP0327-025GEA
22.39
USD
TCP0327-025G
TCP0327-500G
4-(Isopropylamino)diphenylamine ≥98.0% (by HPLC, titration analysis)
4-(Isopropylamino)diphenylamine
CAS Number: 101-72-4
MDL Number: MFCD00026348
Molecular Formula: C15H18N2
Molecular Weight: 226.32
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 180
Melting point (°C): 80
Flash Point (°C): 198
Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: N-Isopropyl-N'-phenyl-1,4-phenylenediamine
Formula:
C₁₅H₁₈N₂ MW: 226.32 g/mol Boiling Pt: 180 °C Melting Pt: 80 °C Flash Pt: 198 °C |
MDL Number:
MFCD00026348 CAS Number: 101-72-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...