Dibutyl phosphate

Supplier: TCI America

Synonyms: Dibutylphosphate, Phosphoric Acid Dibutyl Ester

P0267-500G P0267-25G
TCP0267-500GEA 40.7 USD
TCP0267-500G TCP0267-25G
Dibutyl phosphate
Dibutyl phosphate

CAS Number: 107-66-4
MDL Number: MFCD00015245
Molecular Formula: C8H19O4P
Molecular Weight: 210.21
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 180
Specific Gravity (20/20): 1.13


Other Product Names / Synonyms: Phosphoric Acid Dibutyl Ester

Formula: C₈H₁₉O₄P
MW: 210.21 g/mol
Density: 1.13 (20 °C)
Flash Pt: 180 °C
MDL Number: MFCD00015245
CAS Number: 107-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR