Dibutyl phosphate ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Dibutylphosphate, Phosphoric Acid Dibutyl Ester
P0260-500G
P0260-25G
TCP0260-500GEA
67
USD
TCP0260-500G
TCP0260-25G
Dibutyl phosphate ≥97.0% (by titrimetric analysis)
Dibutyl phosphate
CAS Number: 107-66-4
MDL Number: MFCD00015245
Molecular Formula: C8H19O4P
Molecular Weight: 210.21
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Boiling point (°C): 138
Melting point (°C): -13
Flash Point (°C): 196
Specific Gravity (20/20): 1.06
Other Product Names / Synonyms: Phosphoric Acid Dibutyl Ester
Formula:
C₈H₁₉O₄P MW: 210.21 g/mol Boiling Pt: 138 °C Melting Pt: –13 °C Density: 1.06 (20 °C) Flash Pt: 196 °C |
MDL Number:
MFCD00015245 CAS Number: 107-66-4 |
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...