Cresyl diphenyl phosphate (mixture of analogue) ≥93.0% (so called)
Supplier: TCI America
Synonyms:
Diphenyl p-tolyl phosphate, Diphenyl Cresyl Phosphate, Phosphoric Acid Cresyl Diphenyl Ester
P0259-500ML
P0259-25ML
TCP0259-500MLEA
105.7
USD
TCP0259-500ML
TCP0259-25ML
Cresyl diphenyl phosphate (mixture of analogue) ≥93.0% (so called)
Cresyl diphenyl phosphate (mixture of analogue)
CAS Number: 26444-49-5
MDL Number: MFCD00014920
Molecular Formula: C19H17O4P
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 390
Flash Point (°C): 232
Freezing point (°C): -38
Specific Gravity (20/20): 1.21
Other Product Names / Synonyms: Diphenyl Cresyl Phosphate, Phosphoric Acid Cresyl Diphenyl Ester
Formula:
C₁₉H₁₇O₄P MW: 340.3097 g/mol Boiling Pt: 390 °C Density: 1.21 (20 °C) Flash Pt: 232 °C |
MDL Number:
MFCD00014920 CAS Number: 26444-49-5 |
Specification Test Results
Purity/Analysis Method | >93.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...