p-Phenylenediamine sulfate ≥98.0%
Supplier: TCI America
P0175-500G
P0175-25G
TCP0175-500GEA
67
USD
TCP0175-500G
TCP0175-25G
p-Phenylenediamine sulfate ≥98.0%
p-Phenylenediamine sulphate
CAS Number: 16245-77-5
MDL Number: MFCD00035510
Molecular Formula: C6H8N2
Molecular Weight: 206.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: 1,4-Diaminobenzene Sulfate
Formula:
C₆H₁₀N₂O₄S |
MDL Number:
MFCD00035510 CAS Number: 16245-77-5 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...