N-Phenylurethane ≥98.0%
Supplier: TCI America
P0165-25G
P0165-500G
TCP0165-025GEA
49
USD
TCP0165-025G
TCP0165-500G
N-Phenylurethane ≥98.0%
N-Phenylurethane
CAS Number: 101-99-5
MDL Number: MFCD00026806
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 237
Melting point (°C): 52
Other Product Names / Synonyms: Ethyl Carbanilate, Ethyl N-Phenylcarbamate, N-Phenylcarbamic Acid Ethyl Ester
Formula:
C₉H₁₁NO₂ Boiling Pt: 237 °C Melting Pt: 52 °C |
MDL Number:
MFCD00026806 CAS Number: 101-99-5 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...