D-(-)-Penicillamine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
H-D-Pen-OH, (S)-2-Amino-3-mercapto-3-methylbutanoic acid, Penicillamine
CAS Number: 52-67-5
MDL Number: MFCD00064302
Molecular Formula: C5H11NO2S
Molecular Weight: 149.21
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: -63 deg (C=1, 1mol/L NaOH sol.)
Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Immunosuppressant Ingredients, Anti-rheumatoid Arthritis Ingredients, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: D-beta,beta-Dimethylcysteine, D-beta-Mercaptovaline
Formula:
C₅H₁₁NO₂S MW: 149.2 g/mol |
MDL Number:
MFCD00064302 CAS Number: 52-67-5 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...