cis-5-Octen-1-ol ≥90.0%

Supplier: TCI America

Synonyms: cis-5-Octen-1-ol, (Z)-Oct-5-en-1-ol

O0250-25ML O0250-5ML
TCO0250-25MLEA 281 USD
TCO0250-25ML TCO0250-5ML
cis-5-Octen-1-ol ≥90.0%
cis-5-Octen-1-ol

CAS Number: 64275-73-6
MDL Number: MFCD00015569
Molecular Formula: C8H16O
Molecular Weight: 128.22
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 97
Flash Point (°C): 82
Specific Gravity (20/20): 0.85

Formula: C₈H₁₆O
MW: 128.22 g/mol
Boiling Pt: 97 °C
Density: 0.85 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00015569
CAS Number: 64275-73-6

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR