Octyltrimethylammonium bromide ≥98.0%
Supplier: TCI America
Synonyms:
Trimethyl-1-octylammonium bromide
O0163-500G
O0163-25G
TCO0163-500GEA
442
USD
TCO0163-500G
TCO0163-025G
Octyltrimethylammonium bromide ≥98.0%
Octyltrimethylammonium bromide
CAS Number: 2083-68-3
MDL Number: MFCD00051873
Molecular Formula: C11H26BrN
Molecular Weight: 252.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 217
Product Categories: Materials Chemistry, Cationic Surfactants, Bioscience, Quaternary Ammonium Salts for Clinical Chemistry
Other Product Names / Synonyms: Trimethyl-n-octylammonium Bromide
Formula:
C₁₁H₂₆BrN MW: 252.24 g/mol Melting Pt: 217 °C |
MDL Number:
MFCD00051873 CAS Number: 2083-68-3 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...