7-Nitro-1H-indazole ≥97.0%

Supplier: TCI America

Synonyms: 7-Nitroindazole, 7-NINA, 7-Nitro-1H-indazole

N0827-1G N0827-5G
TCN0827-1GEA 63.8 USD
TCN0827-1G TCN0827-5G
7-Nitro-1H-indazole ≥97.0%
7-Nitro-1H-indazole

CAS Number: 2942-42-9
MDL Number: MFCD00022789
Molecular Formula: C7H5N3O2
Molecular Weight: 163.14
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 186

Formula: C₇H₅N₃O₂
MW: 163.14 g/mol
Melting Pt: 186 °C
MDL Number: MFCD00022789
CAS Number: 2942-42-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d186197eaafa47f2d64f48c23b9038d68ee89a0a&vid=d186197eaafa47f2d64f48c23b9038d68ee89a0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR