2-Nitrophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: Dihydroxy(2-nitrophenyl)borane, (2-Nitrophenyl)boronic acid, 2-Nitrobenzeneboronic acid, (2-Nitrophenyl)metaboric acid

N0811-5G N0811-1G
TCN0811-5GEA 184.1 USD
TCN0811-5G TCN0811-1G
2-Nitrophenylboronic acid (contains varying amounts of Anhydride)
2-Nitrophenylboronic acid

CAS Number: 5570-19-4
MDL Number: MFCD00161358
Molecular Formula: C6H6BNO4
Molecular Weight: 166.93
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 142


Other Product Names / Synonyms: 2-Nitrobenzeneboronic Acid

Formula: C₆H₆BNO₄
MW: 166.93 g/mol
Melting Pt: 142 °C
MDL Number: MFCD00161358
CAS Number: 5570-19-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc328068fc74139da075b8eb8b1d095230223107&vid=cc328068fc74139da075b8eb8b1d095230223107&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR