2-Nitrobenzenesulfonamide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

N0705-25G N0705-5G
TCN0705-25GEA 166 USD
TCN0705-25G TCN0705-5G
2-Nitrobenzenesulfonamide ≥98.0% (by HPLC, titration analysis)
2-Nitrobenzenesulphonamide

CAS Number: 5455-59-4
MDL Number: MFCD00009807
Molecular Formula: C6H6N2O4S
Molecular Weight: 202.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 191

Formula: C₆H₆N₂O₄S
MW: 202.19 g/mol
Melting Pt: 191 °C
MDL Number: MFCD00009807
CAS Number: 5455-59-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4dd3c6d82cec4eaec7f98cf0cc2b0af9cd3e2d5&vid=f4dd3c6d82cec4eaec7f98cf0cc2b0af9cd3e2d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR