1-Naphthylacetonitrile ≥98.0%

Supplier: TCI America

Synonyms: 2-(Naphthalen-1-yl)acetonitrile

N0366-25G N0366-500G
TCN0366-025GEA 85 USD
TCN0366-025G TCN0366-500G
1-Naphthylacetonitrile ≥98.0%
1-Naphthylacetonitrile

CAS Number: 132-75-2
MDL Number: MFCD00004041
Molecular Formula: C12H9N
Molecular Weight: 167.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 113
Freezing point (°C): 29


Other Product Names / Synonyms: 1-Naphthylmethylcyanide

Formula: C₁₂H₉N
MW: 167.21 g/mol
Flash Pt: 113 °C
MDL Number: MFCD00004041
CAS Number: 132-75-2
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32ffb88008216164f1586916b08bafa599d43601&vid=32ffb88008216164f1586916b08bafa599d43601&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR