2-Methoxynicotinic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-Methoxy-3-pyridinecarboxylic acid, 2-Methoxypyridine-3-carboxylic acid
M2041-25G
M2041-5G
TCM2041-25GEA
194
USD
TCM2041-25G
TCM2041-5G
2-Methoxynicotinic acid ≥98.0% (by GC, titration analysis)
2-Methoxynicotinic acid
CAS Number: 16498-81-0
MDL Number: MFCD00085110
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 150
Other Product Names / Synonyms: 2-Methoxy-3-pyridinecarboxylic Acid
Formula:
C₇H₇NO₃ MW: 153.14 g/mol Melting Pt: 150 °C |
MDL Number:
MFCD00085110 CAS Number: 16498-81-0 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...