Methyl-ɑ-bromophenylacetate ≥98.0%
Supplier: TCI America
Synonyms:
Methyl alpha-broMophenylacetate, ɑ-Bromophenylacetic Acid Methyl Ester
M2038-5G
M2038-25G
TCM2038-5GEA
71
USD
TCM2038-5G
TCM2038-25G
Methyl-ɑ-bromophenylacetate ≥98.0%
Methyl-alpha-bromophenylacetate
CAS Number: 3042-81-7
MDL Number: MFCD00013535
Molecular Formula: C9H9BrO2
Molecular Weight: 229.07
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 172
Flash Point (°C): 150
Specific Gravity (20/20): 1.46
Storage Temperature: 0-10°C
Other Product Names / Synonyms: alpha-Bromophenylacetic Acid Methyl Ester
Formula:
C₉H₉BrO₂ MW: 229.07 g/mol Boiling Pt: 172 °C Density: 1.46 (20 °C) Flash Pt: 150 °C |
MDL Number:
MFCD00013535 CAS Number: 3042-81-7 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...