Methyl difluoroacetate ≥97.0%

Supplier: TCI America

M2021-25G M2021-5G
TCM2021-25GEA 105.12 USD
TCM2021-25G TCM2021-5G
Methyl difluoroacetate ≥97.0%
Methyl difluoroacetate

CAS Number: 433-53-4
MDL Number: MFCD00039276
Molecular Formula: C3H4F2O2
Molecular Weight: 110.06
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 86
Flash Point (°C): 24
Specific Gravity (20/20): 1.27


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Difluoroacetic Acid Methyl Ester

Formula: C₃H₄F₂O₂
MW: 110.06 g/mol
Boiling Pt: 86 °C
Density: 1.27 (20 °C)
Flash Pt: 24 °C
MDL Number: MFCD00039276
CAS Number: 433-53-4
UN: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46a7ffd7e9be847b11dcee793e7111417a4d1ace&vid=46a7ffd7e9be847b11dcee793e7111417a4d1ace&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR