(R)-1-Methyl-3-pyrrolidinol ≥98.0%

Supplier: TCI America

Synonyms: (R)-3-Hydroxy-1-methylpyrrolidine, (R)-(-)-1-Methyl-3-pyrrolidinol, (R)-3-Hydroxy-1-methyl-pyrrolidine

M1937-1G M1937-5G
TCM1937-1GEA 96.2 USD
TCM1937-1G TCM1937-5G
(R)-1-Methyl-3-pyrrolidinol ≥98.0%
(R)-1-Methyl-3-pyrrolidinol

CAS Number: 104641-60-3
MDL Number: MFCD03788747
Molecular Formula: C5H11NO
Molecular Weight: 101.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 106
Flash Point (°C): 71
Specific Gravity (20/20): 0.98
Specific rotation [a]20/D: -7 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-3-Hydroxy-1-methylpyrrolidine

Formula: C₅H₁₁NO
Boiling Pt: 106 °C
Density: 0.98 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD03788747
CAS Number: 104641-60-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a77a7ee65c16889c89bdf72fe3a628dcd4540ea&vid=7a77a7ee65c16889c89bdf72fe3a628dcd4540ea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR