6-Methylnicotinic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
6-Methyl-3-pyridinecarboxylic acid, 6-Methylpyridine-3-carboxylic acid
M1894-25G
M1894-5G
TCM1894-25GEA
319
USD
TCM1894-25G
TCM1894-5G
6-Methylnicotinic acid ≥98.0% (by GC, titration analysis)
6-Methylnicotinic acid
CAS Number: 3222-47-7
MDL Number: MFCD00006341
Molecular Formula: C7H7NO2
Molecular Weight: 137.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 210
Other Product Names / Synonyms: 6-Methylpyridine-3-carboxylic Acid
Formula:
C₇H₇NO₂ MW: 137.14 g/mol Melting Pt: 210 °C |
MDL Number:
MFCD00006341 CAS Number: 3222-47-7 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...