L-Glutamic acid 5-methyl ester ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: H-Glu(OMe)-OH

M1690-25G M1690-5G
TCM1690-25GEA 110 USD
TCM1690-25G TCM1690-5G
L-Glutamic acid 5-methyl ester ≥98.0% (by titrimetric analysis)
L-Glutamic acid 5-methyl ester

CAS Number: 1499-55-4
Molecular Formula: C6H11NO4
Molecular Weight: 161.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 177
Specific rotation [a]20/D: 29 deg (C=2, 6mol/L HCl)


Product Categories: Bioscience, Side Chain Protected Amino Acids (N,C-termini Free)
Other Product Names / Synonyms: L-Glutamic Acid 5-Methyl Ester, H-Glu(OMe)-OH

Formula: C₆H₁₁NO₄
MW: 161.2 g/mol
Melting Pt: 177 °C
MDL Number: MFCD00002632
CAS Number: 1499-55-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76979773a6330a20571d5762f2a4720501fe5a06&vid=76979773a6330a20571d5762f2a4720501fe5a06&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR