Morpholinosulfur trifluoride ≥93.0%
Supplier: TCI America
Synonyms:
Morph-DAST
M1573-5G
M1573-1G
TCM1573-5GEA
149
USD
TCM1573-5G
TCM1573-1G
Morpholinosulfur trifluoride ≥93.0%
Morpholinosulfur trifluoride
CAS Number: 51010-74-3
MDL Number: MFCD00037057
Molecular Formula: C4H8F3NOS
Molecular Weight: 175.17
Purity/Analysis Method: >93.0% (T)
Form: Clear Liquid
Boiling point (°C): 42
Flash Point (°C): 76
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Glycosylation, Fluorinating Agents, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: Morph-DAST
Formula:
C₄H₈F₃NOS MW: 175.17 g/mol Boiling Pt: 42 °C Flash Pt: 76 °C |
MDL Number:
MFCD00037057 CAS Number: 51010-74-3 |
|
Specification Test Results
Purity/Analysis Method | >93.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...