3-(4-Methoxybenzoyl)propionic acid ≥98.0%
Supplier: TCI America
Synonyms:
4-(4-Methoxyphenyl)-4-oxobutanoic acid
M1383-25G
M1383-5G
TCM1383-25GEA
175
USD
TCM1383-25G
TCM1383-5G
3-(4-Methoxybenzoyl)propionic acid ≥98.0%
3-(4-Methoxybenzoyl)propionic acid
CAS Number: 3153-44-4
MDL Number: MFCD00002795
Molecular Formula: C11H12O4
Molecular Weight: 208.21
Purity/Analysis Method:
Other Product Names / Synonyms: 4-(4-Methoxyphenyl)-4-oxobutyric Acid
Formula:
C₁₁H₁₂O₄ Melting Pt: 149 °C |
MDL Number:
MFCD00002795 CAS Number: 3153-44-4 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...