5-Methylquinoxaline ≥98.0%
Supplier: TCI America
M1371-5G
M1371-1G
TCM1371-5GEA
95.2
USD
TCM1371-5G
TCM1371-1G
5-Methylquinoxaline ≥98.0%
5-Methylquinoxaline
CAS Number: 13708-12-8
MDL Number: MFCD00012335
Molecular Formula: C9H8N2
Molecular Weight: 144.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 110
Specific Gravity (20/20): 1.13
Formula:
C₉H₈N₂ MW: 144.18 g/mol Boiling Pt: 120 °C Density: 1.13 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00012335 CAS Number: 13708-12-8 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...