Methyl (4-methoxyphenyl)acetate ≥97.0%

Supplier: TCI America

Synonyms: Methyl 4-methoxyphenylacetate

M1330-25G M1330-500G
TCM1330-025GEA 47 USD
TCM1330-025G TCM1330-500G
Methyl (4-methoxyphenyl)acetate ≥97.0%
Methyl (4-methoxyphenyl)acetate

CAS Number: 23786-14-3
MDL Number: MFCD00008454
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 158
Flash Point (°C): 100
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: 4-Methoxyphenylacetic Acid Methyl Ester

Formula: C₁₀H₁₂O₃
MW: 180.23 g/mol
Boiling Pt: 158 °C
Density: 1.12 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00008454
CAS Number: 23786-14-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3437820cc8b341e1a635015664d3fdb669deb65d&vid=3437820cc8b341e1a635015664d3fdb669deb65d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR